Learn More
Heathrow Scientific MagFuge™ High Speed Centrifuge Rotor
Shop All Fisherbrand Products
Click to view available options
Capacity:
12 x 1.5/2 mL
6 x 5 mL
Unit Size:
Each
Specifications
Specifications
| Description | High Speed Centrifuge Rotor |
| Material | Polypropylene |
| Type | Rotor |
| Capacity | 6 x 5 mL |
| Max. RCF | 9,785 x g |
| Max. Speed | 12,500 rpm |
| For Use With (Equipment) | Centrifuge |
| Angle | Angled, Fixed |
| No. of Places | 12 |