missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 539.00
Specifications
| Antigen | MAGI1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18296881
|
Novus Biologicals
NBP2-58071 |
100 μL |
€ 539.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627856
|
Novus Biologicals
NBP2-58071-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGI1 Polyclonal specifically detects MAGI1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MAGI1 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| AIP-3, AIP3WW and PDZ domain-containing protein 1, membrane associated guanylate kinase, WW and PDZ domain containing 1, TNRC19BAP-1, WWP3WW domain-containing protein 3 | |
| MAGI1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 9223 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IVEVNKKNVQALTHNQVVDMLVECPKGSEVTLLVQRGGLPVPKKSPKSQPLERKDSQNSSQHSVSSHRSLHTASPSHSTQVLPEFPPAEAQAPDQTD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title