missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10264580 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10264580

Brand: Fisher Chemical M/0550/65

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C
Color White
pH 5 to 6.5
Physical Form Powder/Solid
Quantity 5 kg
Formula Weight 203.31g/mol
Packaging Plastic keg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.