missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, Mg/Al/Zn, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Magnesium, Mg/Al/Zn, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Lanthanum/Cerium/Nickel/Cadmium/Calcium/Silver/Lead/Silicon/Tin/Manganese/Copper/Zinc/Aluminium/Iron/Beryllium/Mercury |
| CAS | 7439-95-4 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lanthanum 0.0021 %/Cerium 0.0024 %/Nickel 0.0039 %/Cadmium 0.0055 %/Calcium 0.0064 %/Silver 0.0074 %/Lead 0.0085 %/Silicon 0.022 %/Tin 0.043 %/Manganese 0.092 %/Copper 0.125 %/Zinc 0.925 %/Aluminium 7.45 %/Iron (0.008) %/Beryllium (0.01) %/Mercury (0.033) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction