missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium, Mg/Al/Zn, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Magnesium, Mg/Al/Zn, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Lead/Cadmium/Tin/Silver/Iron/Nickel/Calcium/Copper/Zinc/Manganese/Silicon/Aluminium |
| CAS | 7439-95-4 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.0009 %/Cadmium 0.0049 %/Tin 0.005 %/Silver 0.0064 %/Iron 0.0069 %/Nickel 0.0141 %/Calcium 0.021 %/Copper 0.0215 %/Zinc 0.128 %/Manganese 0.203 %/Silicon 0.33 %/Aluminium 4.2 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction