missing translation for 'onlineSavingsMsg'
Learn More

Magnesium, Mg/Mn, 50g, ARMI∣MBH

Product Code. 17992506
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
17992506 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 17992506

Brand: ARMI/MBH MBH63X MGE3 CC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Magnesium, Mg/Mn, 50g
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron/Nickel/Silver/Cadmium/Lead/Tin/Copper/Zinc/Silicon/Manganese/Aluminium/Beryllium
CAS 7439-95-4
Quantity 50 g
Form Pieces
Concentration or Composition (by Analyte or Components) Iron 0.0005 %/Nickel 0.0028 %/Silver 0.0041 %/Cadmium 0.0046 %/Lead 0.0046 %/Tin 0.006 %/Copper 0.0072 %/Zinc 0.0101 %/Silicon 0.014 %/Manganese 1.62 %/Aluminium (0.056) %/Beryllium <0.0001 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.