Learn More
Magnesium Nitrate Hexahydrate, puriss p.a, 98.0 to 102.0% (KT), Honeywell Fluka™
Product Code. 15616070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15616070 250 g Poly bottle 250g
15606070 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15616070 Supplier Honeywell-Fluka Supplier No. 63087250G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Puriss. p.a., ACS Reagent, 98.0-102.0%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Specifications
Melting Point 89°C
Color Colorless
Physical Form Solid
Quantity 250 g
Linear Formula Mg(NO3)2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 256.41g/mol
Percent Purity 98.0 to 102.0% (KT)
Odor Odorless
Packaging Poly bottle
Density 1.464g/cm3; Vapor Density: 6 (vs air)
Chemical Name or Material Magnesium nitrate hexahydrate