missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Oxide (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Magnesium oxide |
| CAS | 1309-48-4 |
| Quantity | 50 g |
| Molecular Formula | Mg O |
| Synonym | Magnesium oxide (MgO), Magnesium oxide (8CI), 1000A, 100A, 100A (oxide), 2000A, 300A, 3310FY, 500-04R, 500A, 995S, 99NC, 99SBS, A 10, A 10S, A 25, A 45, A 45-325th, A 45-500th, AD 100P, AM 2, AM 2 (cement additive), Akro-Mag Bars Green, Akro-Mag Green, Animag, Anscor P, BayMag, BayMag 30, BayMag 40, CX 50, Calcined magnesia, Causmag, Caustic magnesite, D 50, DAB 6, DAB 6 (oxide), Denmag KMAOH-F, E 10, E 10 (oxide), E 4, E 4 (oxide), EMAG 1000, Ekorokku 1000I, ElastoMag 170 Special, Elastomag 100, Elastomag 170, FMR-PC, FNM-G, Fert-O-Mag, FloMag HP, FloMag HP-ER, H 10, H 10 (oxide), HP 10, HP 10 (oxide), HP 10N, HP 30, HP 30 (oxide), HP 30A, HTMg 02, Hamag LP, Insulmag 4, JRC-MGO 3, KM 3, KM 3 (oxide), KM 3150, KM 40, KMA-X, KMAO-H, KMAOH-F, KMB 100-200, KP 3083, KP-JM, KPLL 20, KPLL 60, Keisho Magu 92325, Kyowaad 100, Kyowamag 100, Kyowamag 150, Kyowamag 150-1, Kyowamag 150B, Kyowamag 150C, Kyowamag 20, Kyowamag 3-150, Kyowamag 30, Kyowamag 40, Kyowamag 60, Kyowamag LCM, Kyowamag MF 150, Kyowamag MF 30, Kyowaway 150, LHN 01, Liquimag A, Liquimag B, Luvatol 35, Luvatol DH 25, Luvatol EH 35, Luvatol EK 100KM, Luvatol MK 35, MA 150, MA 150 (oxide), MCP 524-50, MF 150, MF 30, MG 011PB, MG 15, MGO 11PB, MJ 30, MK 35, MM 469, MTK 30, Mag Chem 10, Mag Chem 10-200, Mag Chem 10-325, Mag Chem 10-325S, Mag Chem 10-40, Mag Chem 10CR, Mag Chem 200AD, Mag Chem 200D, Mag Chem 30, Mag Chem 35, Mag Chem 40, Mag Chem 50, Mag Chem 50M, Mag Chem 50Y, Mag Chem P 98, MagOx 54, MagOx 93HR, MagOx 98HR, MagOx 98HR-STI, Magcal, Maglax, Maglite, Maglite 170, Maglite CG 1, Maglite D, Maglite DE, Maglite K, Maglite S, Maglite Y, Magmic, Magmic 4, Magmicron MD 4AM, Magmit, Magnesa Preprata, Magnesia, Magnesia (MgO), Magnesia 100A, Magnesia 2923, Magnesia 312, Magnesia 500A, Magnesia usta, Magnesium monoxide, Magnesium(II) oxide, Magox, Magsarat 150ST, Marmag, Micromag, Micromag 150, Micromag 3-150, Micromag 3-30, NanoActive Magnesium Oxide, NanoActive Magnesium Oxide Plus, NanoActive MgO, NanoActive MgO Plus, Nanotek MgO, Neutran BMR, P-WR 2, PG 9033, PSF-WZ, PurMag 170, Puremag FNM-G, Pyrokisuma 3310S, Pyrokisuma 3320, Pyrokisuma 3320A, Pyrokisuma 3320K, Pyrokisuma 3350, Pyrokisuma 5201, Pyrokisuma 530, Pyrokisuma 5301, Pyrokisuma 5301J, Pyrokisuma 5301K, Pyrokisuma 5Q, Pyrokisuma N, R 150, RA, RA 150, RF 100SC, RF 10C, RF 10C-AC, RF 10C-C, RF 10C-EC, RF 10C-SC, RF 10CS, RF 10SC, RF 20C-HR, RF 30C, RF 50, RF 50AC, RF 50C, RF 50SC, RF 50T, RF 70C, RF 70C-SC, RF 98, RSG, Remag AC, Rf 70, Rhenofit 2120, Rhenofit D/A, SIG, SL, SL-WR 2, SLO 369, SLO 469, SMO, SMO 0.1, SMO 0.4, SMO 02, SMO 05, SMO 1, SMO 2, SMO 5, SMO Large Particle, SMO Small Particle, SMO-S 2, SSP 1, SSP 3, Sammagu Kyowa, Sanka Magnesium EL, Scorchguard O, Starmag 150, Starmag L, Starmag M, Starmag P, Starmag PSF 150, Starmag PSF 150F, Starmag PSF-WR, Starmag SL, Starmag SL-WR, Starmag U, Starmag U 2, T 22, T 22 (oxide), T-TEK, TNM 1, TNM 21, TTA, TTA (metal oxide), Tatehomag 500, Tomex AD 100, Tomita AD 100P, U 10, U 180P, UBE 995S, UC 95H, UC 95M, UC 95S, UC 99, UCM 150, Ube 100A, Ube 500A, Ube 98HD, Ube Green, VK-Mg 30, Zabotan Mg, Zolitho 40, magnesium oxide 75 |
| InChI Formula | InChI=1S/Mg.O |
| SMILES | O=[Mg] |
| IUPAC Name | oxomagnesium |
| Molecular Weight (g/mol) | 40.3 |
| Formula Weight | 39.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?