Learn More
Velp Scientifica™ Magnetic Stirring Bars
Product Code. p-8014165 Shop All Velp Products
Click to view available options
Length (Metric):
6 mm
12 mm
60 mm
Diameter (Metric):
3 mm
4.5 mm
10 mm
Unit Size:
Each
Product Code. 15329514
Brand: Velp Scientifica™ A00001062
Only null left
Add to basket
Add to basket
Emerse this into a liquid to produce a stirring movement. Velp Scientifica™ Magnetic Stirring Bars do not contaminate or react with the mixure they are in.
  • For stirring applications
  • Chemically inert
Specifications
For Use With (Equipment) VELP Heating magnetic stirrer with round top
Diameter (Metric) 3 mm
Length (Metric) 6 mm
Type Stir Bar