missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Majors, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Brass Majors, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Chromium/Selenium/Silver/Cobalt/Arsenic/Silicon/Antimony/Bismuth/Lead/Phosphorus/Tin/Iron/Aluminium/Manganese/Nickel/Zinc/Copper |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0005 %/Chromium 0.0006 %/Selenium 0.0009 %/Silver 0.003 %/Cobalt 0.0062 %/Arsenic 0.0099 %/Silicon 0.012 %/Antimony 0.0207 %/Bismuth 0.021 %/Lead 0.022 %/Phosphorus 0.033 %/Tin 0.047 %/Iron 0.135 %/Aluminium 0.149 %/Manganese 0.16 %/Nickel 1.56 %/Zinc 36.6 %/Copper 60.9 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction