missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML3 Mouse, Clone: MAML3/1303, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 55534-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
MAML3 | |
Cytoplasmic | |
MAML3/1303 | |
Unconjugated | |
NP_061187 | |
Mouse | |
150-170kDa | |
Protein A/G Purified | |
RUO | |
55534 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
MAML3 | |
Recombinant human full-length MAML3 protein | |
MCF-7 cells. Pancreas or Placenta. Cervix or Colon Carcinoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |