Learn More
MAN2A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92006-0.02ml
This item is not returnable.
View return policy
Description
MAN2A1 Polyclonal antibody specifically detects MAN2A1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MAN2A1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| Alpha-mannosidase 2;AMan II;GOLIM7;MANA2;MANII | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 865-1144 of human MAN2A1 (NP_002363.2). SNIVDIRKVYNREIAMKISSDIKSQNRFYTDLNGYQIQPRMTLSKLPLQANVYPMTTMAYIQDAKHRLTLLSAQSLGVSSLNSGQIEVIMDRRLMQDDNRGLEQGIQDNKITANLFRILLEKRSAVNTEEEKKSVSYPSLLSHITSSLMNHPVIPMANKFSSPTLELQGEFSPLQSSLPCDIHLVNLRTIQSKVGNGHSNEAALILHRKGFDCRFSSKGTGLFCSTTQGKILVQKLLNKFIVESLTPSSLSLMHSPPGTQNISEINLSPMEISTFRIQLR | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 4124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |