missing translation for 'onlineSavingsMsg'
Learn More

MAN2A1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 204.00 - € 463.00

Specifications

Antigen MAN2A1
Dilution Western Blot 1:1000-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18662721
View Documents
Novus Biologicals
NBP2-92006-0.02ml
0.02 mL
€ 204.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18610661
View Documents
Novus Biologicals
NBP2-92006-0.1ml
0.1 mL
€ 463.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MAN2A1 Polyclonal antibody specifically detects MAN2A1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

MAN2A1
Western Blot
Unconjugated
Rabbit
Cancer, Endocrinology, Signal Transduction
PBS with 50% glycerol, pH7.3.
4124
IgG
Affinity purified
Western Blot 1:1000-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
Alpha-mannosidase 2;AMan II;GOLIM7;MANA2;MANII
Recombinant fusion protein containing a sequence corresponding to amino acids 865-1144 of human MAN2A1 (NP_002363.2). SNIVDIRKVYNREIAMKISSDIKSQNRFYTDLNGYQIQPRMTLSKLPLQANVYPMTTMAYIQDAKHRLTLLSAQSLGVSSLNSGQIEVIMDRRLMQDDNRGLEQGIQDNKITANLFRILLEKRSAVNTEEEKKSVSYPSLLSHITSSLMNHPVIPMANKFSSPTLELQGEFSPLQSSLPCDIHLVNLRTIQSKVGNGHSNEAALILHRKGFDCRFSSKGTGLFCSTTQGKILVQKLLNKFIVESLTPSSLSLMHSPPGTQNISEINLSPMEISTFRIQLR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.