Learn More
Manganese(II) Sulfate Monohydrate, ≥99%, Honeywell™, Reagentplus™
Product Code. p-7120547 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
100g
1kg
500g
Product Code. 15667960
Brand: Honeywell Research Chemicals M7634100G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reagent Grade,≥99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O
Specifications
Melting Point 700°C
Color Red
pH 3.0 to 3.5 (at 50g/L 20°C)
Physical Form Powder
Quantity 100 g
Linear Formula MnSO4 · H2O
UN Number UN3077
Formula Weight 169.02g/mol
Percent Purity ≥99%
Packaging Poly bottle
Chemical Name or Material Manganese(II) sulfate monohydrate