Learn More
B Braun™ Manufix™ Sensitive Natural Latex Exam Gloves
Product Code. p-8005420
Click to view available options
Size:
Small
Large
Unit Size:
Pack of 100
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10742122
Brand: B Braun™ 9209484
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Manufactured from powder-free natural rubber latex. B Braun™ Manufix™ Sensitive Natural Latex Exam Gloves are non-sterile, disposable, and suitable for use with food.
  • Manufactured from natural rubber latex
  • Single-use
  • Powder-free
  • Non-sterile
  • Suitable for use with food
Specifications
Color Natural White
Disposable Single-use
Exterior Finish Micro-rough
Material Rubber Latex
Powdered Powder Free
Product Type Examination Gloves
Size Large
Thickness (Metric) ≥0.11 mm
Finger Style Textured
For Use With (Application) Suitable for food processing
Glove Cuff Style Rolled
Quantity 100 Pk.
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato