missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21633-1-AP-150UL
This item is not returnable.
View return policy
Description
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.Specifications
| MAP1B | |
| Polyclonal | |
| Unconjugated | |
| MAP1B | |
| MAP 1B, MAP1B, MAP5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17755, 4131 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14873, P46821 | |
| Map1b | |
| MAP1B Fusion Protein Ag16255 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction