missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MAP1B |
|---|---|
| Concentration | 0.49 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16851235
|
Proteintech
21633-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16841235
|
Proteintech
21633-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.Specifications
| MAP1B | |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P14873, P46821 | |
| 17755, 4131 | |
| MAP1B Fusion Protein Ag16255 | |
| Primary | |
| -20°C | |
| MAP1B |
| 0.49 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| MAP 1B, MAP1B, MAP5 | |
| Map1b | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title