missing translation for 'onlineSavingsMsg'
Learn More

MARCKS Antibody, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-05539-100ul

 View more versions of this product

Product Code. 18642907

  • € 481.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MARCKS Monoclonal antibody specifically detects MARCKS in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MARCKS
Monoclonal
Unconjugated
50% PBS, 50% glycerol
Mouse
Immunogen affinity purified
RUO
Primary
Human, Monkey
Purified
Western Blot, Immunohistochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:2000
80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain
Full length human MARCKS expressed in and purified from E. coli.
100 μg
Lipid and Metabolism, Signal Transduction
4082
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.