Learn More
MARK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84848
This item is not returnable.
View return policy
Description
MARK2 Polyclonal specifically detects MARK2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| EC 2.7.11, EC 2.7.11.1, ELKL motif kinase, ELKL motif kinase 1, EMK-1, EMK1PAR-1, MAP/microtubule affinity-regulating kinase 2MGC99619, PAR1 homolog, Par1b, Ser/Thr protein kinase PAR-1B, serine/threonine protein kinase EMK, serine/threonine-protein kinase MARK2 | |
| RUO |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| 0.1 mL | |
| IgG |