missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody
Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.
€ 118.00 - € 535.00
Specifications
Antigen | MART-1/Melan-A (Melanoma Marker) Ab-4 |
---|---|
Clone | A103 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12612327
|
Epredia™
MS-799-P0 |
Concentrated | 200μg/mL | 100 μL |
€ 118.00
100µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12622327
|
Epredia™
MS-799-P1 |
Concentrated | 200μg/mL | 500 μL |
€ 352.00
500µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12602327
|
Epredia™
MS-799-P |
Concentrated | 200μg/mL | 1 mL |
€ 535.00
1mL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12682327
|
Epredia™
MS-799-R7 |
Prediluted | — | 7 mL |
€ 305.00
7mL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12692327
|
Epredia™
MS-799-RQ |
Ready-to-Use | — | 12 mL |
€ 367.00
12mL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12632327
|
Epredia™
MS-799-P1ABX |
Concentrated | 1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12642327
|
Epredia™
MS-799-PABX |
Concentrated | 1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. MART-1 is present in melanosomes and endoplasmic reticulum.Host Species: Mouse
Clone: A103
Isotype: IgG1
Species Reactivity: Human and Dog. Others not tested.
Epitope: Not determined
Immunogen: Recombinant MART-1 protein
Molecular Weight: 18kDa
Positive Control: CAC1 melanoma cells and Melanomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Specifications
MART-1/Melan-A (Melanoma Marker) Ab-4 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
IVD | |
Recombinant MART-1 protein |
A103 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Primary |