Learn More
Masterflex™ L/S™ End Bell Assembly
Complete your pump head assembly with two end bell assemblies. MasterFlex™ I/P™ End Bell Assemblies are designed for use with Masterflex™ I/P™ 73 Standard Pump Heads.
Brand:  Masterflex™ 07016-91
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11895202
  • € 145.00 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • Assemblies are made from high-performance Polyphenylene sulfide and stainless steel
Specifications
Specifications
For Masterflex L/S Standard 07016-10/–21 pump heads
1 Each
End Bell Assembly