Learn More
MBL™ Graduated Class AS Type 1 Glass Pipets
Product Code. 11389653
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
Color:
Black
Green
Orange
Red
Yellow
Tolerance:
±0.007 mL
±0.01 mL
±0.03 mL
±0.050 mL
±0.10 mL
Unit Size:
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Color Tolerance unitSize
11389653 10 mL 0.1 mL Orange ±0.050 mL Pack of 2
11399653 25 mL 0.1 mL Green ±0.10 mL Pack of 2
11359653 1 mL 0.01 mL Yellow ±0.007 mL Pack of 2
11369653 2 mL 0.02 mL Black ±0.01 mL Pack of 2
11379653 5 mL 0.05 mL Red ±0.03 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11389653 Supplier MBL International Supplier No. PRA776AS
Add to basket
Add to basket
Pipet with precision. MBL™ class AS type 1 soda-lime glass pipets have graduations in permanent amber stain. Type 1 pipets have the zero mark at the top of the pipet.
  • Complies with ISO 835
  • Calibrated to deliver from zero at the top to any graduation line down to the shoulder
  • Graduations in permanent amber stain which resists aggressive washing solutions
  • Each pipette is calibrated to deliver with a +5s waiting time
  • Manufactured from soda lime glass with tooled jets for extra strength and to ensure correct outflow times
TRUSTED_SUSTAINABILITY
Specifications
Calibrated Yes
Certifications/Compliance ISO 835
Material Soda Lime Glass
Graduated Yes
Graduations 0.1 mL
Tolerance ±0.050 mL
Color Orange
Volume (Metric) 10 mL
Type Pipet