missing translation for 'onlineSavingsMsg'
Learn More

MDC1 Antibody (P2B11), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-12890-0.025mg

 View more versions of this product

Product Code. 18723384

  • € 179.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MDC1 Monoclonal specifically detects MDC1 in Human, Mouse, Bovine, Non-human Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MDC1
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
MDC1
GST-tagged recombinant protein corresponding to mouse MDC1 at and around the N-terminus
0.025 mg
Primary
Detects approx 184kDa. This antibody recognizes MDC1 at and around the N-terminus.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence
P2B11
Unconjugated
DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1
Mouse
Protein G purified
RUO
9656
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only