Learn More
MDM2/HDM2, p Ser186 Antibody (2G2 2.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50242
 View more versions of this product
Product Code. 18674438
  • € 372.00 / 0.10mL
Description
MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MDM2/HDM2
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Immunoprecipitation
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Peptide QRKRHKS(p)DSIS. S(p) represent a phosphorylated serine. This peptide represents the portion of MDM2/HDM2 around Serine 186
0.1 mL
Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Tumor Suppressors
4193
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunoprecipitation
2G2 2.2
Unconjugated
PBS
Mouse
Protein A purified