missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2/HDM2, p Ser186 Antibody (2G2 2.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 372.00
Specifications
| Antigen | MDM2/HDM2 |
|---|---|
| Clone | 2G2 2.2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunoprecipitation |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622288
|
Novus Biologicals
NBP2-50242-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674438
|
Novus Biologicals
NBP2-50242 |
0.1 mL |
€ 372.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| MDM2/HDM2 | |
| 1 mg/mL | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Tumor Suppressors | |
| PBS | |
| 4193 | |
| IgG1 | |
| Protein A purified |
| 2G2 2.2 | |
| Western Blot 1:100 - 1:2000, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| Peptide QRKRHKS(p)DSIS. S(p) represent a phosphorylated serine. This peptide represents the portion of MDM2/HDM2 around Serine 186 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title