missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1/ABCB1 Antibody (MRK16) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52701-0.1mg
This item is not returnable.
View return policy
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF-ready.
Specifications
| MDR1/ABCB1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| ABCB1 | |
| Drug resistant K-562 cells. | |
| 0.1 mg | |
| Cancer, Membrane Trafficking and Chaperones | |
| 5243 | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF | |
| MRK16 | |
| Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a discontinuous extracellular epitope on P-glycoprotein. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction