missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS1 Antibody (OTI1B4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
€ 256.00 - € 550.00
Specifications
| Antigen | MEIS1 |
|---|---|
| Clone | OTI1B4 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:2000 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306161
|
Novus Biologicals
NBP3-44993-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306669
|
Novus Biologicals
NBP3-44993-30ul |
30 μL |
€ 256.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| MEIS1 | |
| Western Blot 1:2000, Immunohistochemistry 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 4211 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI1B4 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse) | |
| Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1(NP_002389) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title