missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melphalan-modified DNA Antibody (MP5/73) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50504-0.025ml
This item is not returnable.
View return policy
Description
Melphalan-modified DNA Monoclonal antibody specifically detects Melphalan-modified DNA in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry
Specifications
| Melphalan-modified DNA | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| ELISA, Immunohistochemistry | |
| MP5/73 | |
| Unconjugated | |
| PBS | |
| Native DNA reacted with melphalan | |
| 0.025 mL | |
| Cancer | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction