missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melphalan-modified DNA Antibody (MP5/73) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 214.00 - € 372.00
Specifications
| Antigen | Melphalan-modified DNA |
|---|---|
| Clone | MP5/73 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622638
|
Novus Biologicals
NBP2-50504-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659286
|
Novus Biologicals
NBP2-50504 |
0.1 mL |
€ 372.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melphalan-modified DNA Monoclonal antibody specifically detects Melphalan-modified DNA in Nonspecies specific samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Melphalan-modified DNA | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Cancer | |
| PBS | |
| IgG2b | |
| Protein G purified |
| MP5/73 | |
| ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| Native DNA reacted with melphalan | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title