missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mesopram
GREENER_CHOICE

Product Code. 18034522
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18034522

Brand: Tocris Bioscience™ 3053/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Orally active PDE4 inhibitor

Orally active phosphodiesterase (PDE) 4 inhibitor. Inhibits Th1 cell proliferation and decreases production of IFN-γ, TNF-α, IL-10 and iNOS in vitro. Triggers ovulation and exhibits efficacy against experimental autoimmune encephalomyelitis and murine colitis in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189940-24-7
Molecular Formula C14H19NO4

Specifications

Quantity 50 mg
Formula Weight 265.31
Purity >98%
Molecular Weight (g/mol) 265.309
Chemical Name or Material Mesopram
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.