missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Nucleoprotein Antibody (hMPV123) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53104-0.025ml
This item is not returnable.
View return policy
Description
Metapneumovirus Nucleoprotein Monoclonal specifically detects Metapneumovirus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.
Specifications
| Metapneumovirus Nucleoprotein | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.02% Sodium Azide | |
| Human MPV strain NCL-145 | |
| 0.025 mL | |
| Virology Bacteria and Parasites | |
| Identification and location of human metapneumovirus (hMPV) of both types A and B. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| hMPV123 | |
| Unconjugated | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction