missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Nucleoprotein Antibody (hMPV123) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.60 - € 416.85
Specifications
| Antigen | Metapneumovirus Nucleoprotein |
|---|---|
| Clone | hMPV123 |
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18081716
|
Novus Biologicals
NBP2-53104 |
0.1 mL |
€ 441.00 € 416.85 / 0.10mL Save € 24.15 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631408
|
Novus Biologicals
NBP2-53104-0.025ml |
0.025 mL |
€ 214.00 € 201.60 / 0.03mL Save € 12.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Metapneumovirus Nucleoprotein Monoclonal specifically detects Metapneumovirus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
| Metapneumovirus Nucleoprotein | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Human MPV strain NCL-145 | |
| Primary | |
| Identification and location of human metapneumovirus (hMPV) of both types A and B. |
| hMPV123 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| IgG2a | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title