missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methylbenzo[b]thiophene-2-carboxylate, 97%

Product Code. 11491057
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11491057

Brand: Thermo Scientific Alfa Aesar B24069.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3133-81-1 | C11H10O2S | 206.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3133-81-1
Molecular Formula C11H10O2S
Molecular Weight (g/mol) 206.26
MDL Number MFCD02683110
InChI Key FDBXIMKCWBPTDG-UHFFFAOYSA-N
Synonym methyl 3-methylbenzo b thiophene-2-carboxylate, methyl 3-methylbenzothiophene-2-carboxylate, 3-methyl-benzo b thiophene-2-carboxylic acid methyl ester, benzo b thiophene-2-carboxylic acid, 3-methyl-, methyl ester, acmc-1cqnl, methyl 3-methylthianaphthene-2-carboxylate, methyl 3-methylbenzo b thiophen-2-carboxylate, 3-methylbenzo b thiophene-2-carboxylic acid methyl ester, benzo b thiophene-2-carboxylicacid, 3-methyl-, methyl ester
PubChem CID 268658
IUPAC Name methyl 3-methyl-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=C(C)C2=CC=CC=C2S1

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
Formula Weight 206.26
Percent Purity 97%
Chemical Name or Material Methyl 3-methylbenzo[b]thiophene-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.