Learn More
Methyl 3-methylbenzo[b]thiophene-2-carboxylate, 97%
Product Code. 11471057
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471057 1 g 1g
11481057 5 g 5g
11491057 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11471057 Supplier Thermo Scientific Alfa Aesar Supplier No. B24069.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 3133-81-1 | C11H10O2S | 206.26 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3133-81-1
Molecular Formula C11H10O2S
Molecular Weight (g/mol) 206.26
MDL Number MFCD02683110
InChI Key FDBXIMKCWBPTDG-UHFFFAOYSA-N
Synonym methyl 3-methylbenzo b thiophene-2-carboxylate, methyl 3-methylbenzothiophene-2-carboxylate, 3-methyl-benzo b thiophene-2-carboxylic acid methyl ester, benzo b thiophene-2-carboxylic acid, 3-methyl-, methyl ester, acmc-1cqnl, methyl 3-methylthianaphthene-2-carboxylate, methyl 3-methylbenzo b thiophen-2-carboxylate, 3-methylbenzo b thiophene-2-carboxylic acid methyl ester, benzo b thiophene-2-carboxylicacid, 3-methyl-, methyl ester
PubChem CID 268658
IUPAC Name methyl 3-methyl-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=C(C)C2=CC=CC=C2S1
Specifications
Melting Point 102°C to 105°C
Quantity 1 g
Formula Weight 206.26
Percent Purity 97%
Chemical Name or Material Methyl 3-methylbenzo[b]thiophene-2-carboxylate