Learn More
Methyl acetopyruvate, 98%, Thermo Scientific™
Product Code. 11433494
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11433494
Brand: Thermo Scientific Alfa Aesar A11190.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20577-61-1
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00043638
InChI Key OMHOEQINEXASKE-UHFFFAOYSA-N
Synonym methyl acetopyruvate, methyl acetylpyruvate, pentanoic acid, 2,4-dioxo-, methyl ester, 2,4-dioxopentanoic acid methyl, 2,4-dioxo-pentanoic acid methyl ester, pentanoic acid,2,4-dioxo-,methyl ester
PubChem CID 88600
IUPAC Name methyl 2,4-dioxopentanoate
SMILES CC(=O)CC(=O)C(=O)OC
Specifications
Melting Point 60°C to 63°C
Quantity 5 g
Formula Weight 144.13
Percent Purity 98%
Chemical Name or Material Methyl acetopyruvate