Learn More
Methyl (R)-(-)-mandelate, 99%
Product Code. 11338945
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338945 5 g 5g
11328945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11338945 Supplier Thermo Scientific Alfa Aesar Supplier No. L14065.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 20698-91-3 | C9H10O3 | 166.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20698-91-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00064247
InChI Key ITATYELQCJRCCK-MRVPVSSYSA-N
Synonym methyl r---mandelate, r---methyl mandelate, r-methyl 2-hydroxy-2-phenylacetate, d---mandelic acid methyl ester, methyl 2r-2-hydroxy-2-phenylacetate, r-methyl mandelate, methyl d---mandelate, r-mandelic acid methyl ester, r---mandelic acid methyl ester, methyl hydroxy phenyl acetate #
PubChem CID 2724623
IUPAC Name methyl (2R)-2-hydroxy-2-phenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)O
Specifications
Melting Point 56°C to 58°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 3589447
Optical Rotation −144° (c=1 in Methanol)
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Methyl (R)-(-)-mandelate