Learn More
Methylglyoxal Antibody (9E7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59368
 View more versions of this product
Product Code. 18061376
  • ONLINE EXCLUSIVE PRICE
    € 679.00
    € 641.55 / 100µg
    Save € 37.45
    5% Off
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Methylglyoxal
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
2-oxopropanal, Pyruvaldehyde
Mouse
Protein G purified
RUO
Primary
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Store at -20C.
IgG2a
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
90000000
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
GLO1
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
100 μg
Cancer
2739
Human, Non-species specific