Learn More
METTL1 Antibody (02), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06375-100ul
 View more versions of this product
Product Code. 18622167
  • € 326.00 / 100µL
Description
METTL1 Monoclonal antibody specifically detects METTL1 in Human samples. It is validated for ELISA, Sandwich ELISA Capture
TRUSTED_SUSTAINABILITY
Specifications
Specifications
METTL1
Monoclonal
Unconjugated
C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human METTL1 (Accession#: NP_005362.3; Asp32-Gln265).
100 μg
Signal Transduction
4234
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
ELISA, Sandwich ELISA Capture
02
0.2 um filtered solution in PBS
Mouse
Protein A purified