missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Antibody (02), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 326.00
Specifications
| Antigen | METTL1 |
|---|---|
| Clone | 02 |
| Applications | ELISA, Sandwich ELISA Capture |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622167
|
Novus Biologicals
NBP3-06375-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674247
|
Novus Biologicals
NBP3-06375-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
METTL1 Monoclonal antibody specifically detects METTL1 in Human samples. It is validated for ELISA, Sandwich ELISA CaptureSpecifications
| METTL1 | |
| ELISA, Sandwich ELISA Capture | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 4234 | |
| IgG | |
| Protein A purified |
| 02 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human METTL1 (Accession#: NP_005362.3; Asp32-Gln265). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title