Learn More
Mgea5 Recombinant Rabbit Monoclonal Antibody (23GB2270), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen MA552491
 View more versions of this product
Product Code. 30088709
  • € 186.00 / 20µL
Description
Mgea5 Recombinant Monoclonal antibody specifically detects Mgea5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mgea5
Recombinant Monoclonal
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O60502, Q8VIJ5, Q9EQQ9
MGEA5, Oga
A synthesized peptide derived from human MGEA5 (2-63AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2270
Unconjugated
Oga
2810009A20Rik; 4833427O07Rik; 5830447M11Rik; AA408215; beta-hexosaminidase; Beta-N-acetylglucosaminidase; beta-N-acetylhexosaminidase; Bifunctional protein NCOAT; Hexc; Hy5; hyaluronidase in meningioma; KIAA0679; MEA5; meningioma expressed antigen 5 (hyaluronidase); meningioma-expressed antigen 5; Mgea5; mKIAA0679; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; NCOAT; Nuclear cytoplasmic O-GlcNAcase and acetyltransferase; OGA; O-GLCNACASE; Protein O-GlcNAcase
Rabbit
Affinity Chromatography
RUO
10724, 154968, 76055
-20°C