missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5 Antibody (1A11D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61828
This item is not returnable.
View return policy
Description
mGluR5 Monoclonal specifically detects mGluR5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| mGluR5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 | |
| Mouse | |
| 132.5 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1A11D10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRM5 | |
| Purified recombinant fragment of human GRM5 (AA: extra 458-580) expressed in E. Coli. | |
| Protein G purified | |
| GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neurotransmission | |
| 2915 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction