missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5 Antibody (1A11D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 217.00 - € 552.00
Specifications
| Antigen | mGluR5 |
|---|---|
| Clone | 1A11D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18291753
|
Novus Biologicals
NBP2-61828 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624727
|
Novus Biologicals
NBP2-61828-0.025ml |
0.025 mL |
€ 217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mGluR5 Monoclonal specifically detects mGluR5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| mGluR5 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neurotransmission | |
| 2915 | |
| Purified recombinant fragment of human GRM5 (AA: extra 458-580) expressed in E. Coli. | |
| Primary | |
| 132.5 kDa |
| 1A11D10 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 | |
| GRM5 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title