missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (AP-MAB0874) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06696-0.1mg
This item is not returnable.
View return policy
Description
MHC Class II Monoclonal specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.
Specifications
| MHC Class II | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| P28078 | |
| HLA-DMA | |
| Activated C57BL/6 mouse spleen cells | |
| 0.1 mg | |
| Immunology, Major Histocompatibility Complexes | |
| 3108 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| AP-MAB0874 | |
| Unconjugated | |
| PBS with No Preservative | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat) immunized with Activated C57BL/6 mouse spleen cells. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction