missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (AP-MAB0874) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 165.00
Specifications
| Antigen | MHC Class II |
|---|---|
| Clone | AP-MAB0874 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready |
| Applications | Flow Cytometry |
Description
MHC Class II Monoclonal specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.Specifications
| MHC Class II | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Immunology, Major Histocompatibility Complexes | |
| PBS with No Preservative | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| HLA-DMA | |
| IgG2 | |
| Protein A or G purified | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat) immunized with Activated C57BL/6 mouse spleen cells. |
| AP-MAB0874 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P28078 | |
| 3108 | |
| Activated C57BL/6 mouse spleen cells | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title