Learn More
Ansell MICRO-TOUCH Nitrile Acc-Free
Product Code. p-200080407 Shop All Ansell Products
Click to view available options
Size:
Large, 8.5/9
X-Small, 5.5/6
Unit Size:
Pack of 100
Product Code. 16346901
Brand: Ansell 700101
Only null left
Add to basket
Add to basket
MICRO-TOUCH Nitrile Acc-Freeare nitrile, powder-free examination gloves with advanced allergy prevention.
  • Superior barrier protection and puncture resistance
  • Chlorinated inner surface for ease of donning
  • Protects from latex Type I allergy and minimizes chemical Type IV allergy
Specifications
Certifications/Compliance (Europe) CE 0493, Category III, Medical Device Regulation (EU) 2017/745 Class I, EN 16523-1, EN 421:2010, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10993-10:2002, ISO 9001, ISO 13485, UKCA
Chemotherapeutic Rated Yes, in accordance with ASTM D6978 (Not listed in the US FDA 510k)
Chlorinated Yes
Accelerator Free Yes
Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4.7 mil
Finger Thickness (Metric) 0.12 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small, 5.5/6
Sterility Non-sterile
Thickness (English) 3.1 mil
Thickness (Metric) 0.08 mm
Length (English) 9.5 in.
Length (Metric) 240 mm