Learn More
Sartorius Stedim Biotech™ Microsart™ RESEARCH Bacteria
Shop All Sartorius Products
Click to view available options
No. of Reactions:
Reagents for 25 PCR reactions
Quantity:
25
Unit Size:
Pack of 25
Description
- Rapidly detect contamination of more than 95 % of all known bacterial species within 2½ hours.
- In a Real-time PCR highly specific TaqMan™ probes bind to a very conserved DNA-sequence that is common for all bacteria (gram-positive and gram negative).
- This kit is designed for contamination control in academia or during drug development and in-process in biopharma or regenerative medicine.
Specifications
Specifications
| Detection Method | qPCR |
| Quantity | 25 |
| No. of Reactions | Reagents for 25 PCR reactions |
| Volume | 2μL |