Learn More
Fisherbrand™ Stainless Steel Straight Tongs
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
410 mm
Unit Size:
Each
Description
- Laboratory straight tongs
- Stainless steel 304 spec 18/8 non-magnetic, bright finish
- Designed for corrosion resistance and durability ideal in laboratory routines
- Laboratory metalware consumables
- Finest in corrosion-resistant 304 stainless steel
- Provides corrosion resistance that prevents product contamination
Specifications
Specifications
| Quantity | 5 Pk. |
| Tip Style | Straight |
| Length (Metric) | 410 mm |
| Material | Mild Steel |
| Type | Laboratory Tongs |