missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIS RII/AMHR2 Antibody (SR1978), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22158-100ul
This item is not returnable.
View return policy
Description
MIS RII/AMHR2 Monoclonal antibody specifically detects MIS RII/AMHR2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MIS RII/AMHR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1978 | |
| Western Blot 1:500-1:2000 | |
| AMH type II receptor, AMHREC 2.7.11.30, MISR2, MISRIIMIS type II receptor, MRII, Muellerian hormone type II receptor, Muellerian hormone type-2 receptor, Mullerian hormone receptor, type II, Mullerian inhibiting substance type II receptor | |
| A synthesized peptide derived from human MIS RII/AMHR2 (Uniprot #: Q16671) | |
| 100 μg | |
| Protein Kinase | |
| 269 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction