missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MIS RII/AMHR2 Antibody (SR1978), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Tekniske data
| Antigen | MIS RII/AMHR2 |
|---|---|
| Clone | SR1978 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18655724
|
Novus Biologicals
NBP3-22158-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689913
|
Novus Biologicals
NBP3-22158-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
MIS RII/AMHR2 Monoclonal antibody specifically detects MIS RII/AMHR2 in Human, Mouse samples. It is validated for Western BlotTekniske data
| MIS RII/AMHR2 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| AMH type II receptor, AMHREC 2.7.11.30, MISR2, MISRIIMIS type II receptor, MRII, Muellerian hormone type II receptor, Muellerian hormone type-2 receptor, Mullerian hormone receptor, type II, Mullerian inhibiting substance type II receptor | |
| A synthesized peptide derived from human MIS RII/AMHR2 (Uniprot #: Q16671) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1978 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 269 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel