Learn More
Invitrogen™ Mitofilin Monoclonal Antibody (K92014_1F4), Invitrogen™
Mouse Monoclonal Antibody
€ 524.00 - € 876.00
Specifications
| Antigen | Mitofilin |
|---|---|
| Clone | K92014_1F4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30212697
|
Invitrogen™
MA555883 |
50 μL |
€ 524.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30212489
|
Invitrogen™
MA556263 |
200 μL |
€ 876.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: FRESVEKTIP YSDKLFEMVL GPAAYNVPLP KKSIQSGPLK ISSVSEVMKE SKQPASQLQK QKGDTPASAT APTEAAQIIS AAGDTLSVPA PAVQPEESLK TDHPEIGEGK PTPALSEEAS SSSIRERPPE EVAARLAQQE KQEQVKIESL AKSLEDALRQ TASVTLQAIA AQNAAVQAVN AHSNILKAAM DNSEIAGEKK SAQWRTVEGA LKERRKAVDE AADALLKAKE ELEKMKSVIE
Mitofilin Monoclonal antibody specifically detects Mitofilin in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| Mitofilin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1700082C19Rik; cell proliferation-inducing gene 4/52 protein; cell proliferation-inducing protein 52; D830041H16Rik; EMBL:AAI05842.1, ECO:0000312; EMBL:AAI05842.1}; HMP; Immt; immt {ECO:0000312; IMMT antibody; inner membrane mitochondrial protein; inner membrane protein, mitochondrial; inner membrane protein, mitochondrial (mitofilin); MGC133830 protein; MIC60; MICOS complex subunit Mic60; MINOS2; mitochondrial inner membrane organizing system 2; mitochondrial inner membrane protein; mitochondrial inner membrane protein {ECO:0000250; Mitofilin; mitofilin {ECO:0000250; motor protein; P87; p87/89; p87/89 antibody; P89; PIG4; PIG52; proliferation-inducing gene 4; RGD:1310684}; UniProtKB:Q8CAQ8, ECO:0000312; UniProtKB:Q8CAQ8} | |
| Recombinant human IMMT (immunogen range is 71-310). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IMMT |
| K92014_1F4 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q16891 | |
| 10989 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.